Learn More
MSC PIP Seamless Knit Gloves, Cut A2, Nitrile Coated, 1 Pair
Catalog No. 5000794294
Encompass
These G-Tek CR seamless knit gloves are designed for cut, puncture, and abrasion resistance, making them suitable for demanding industrial applications. They feature a seamless knit construction for comfort and breathability, and a double-dipped nitrile micro-surface coating on the palm and fingers for enhanced grip in dry and oily conditions.
  • Seamless knit construction offers increased comfort and breathability
  • HPPE blended shell is lightweight, providing excellent dexterity, tactile sensitivity, and cut resistance (ANSI A2)
  • Double-dipped nitrile micro-surface coating provides an elevated grip in dry and oily conditions
  • Coating infused with tiny suction cup pockets that create a vacuum effect, dispensing fluids away on contact
  • Ideal for electronics, glass cutting, sheet metal handling, general duty, parts assembly, and sharp small parts handling
Catalog No. 50-007-94294 Supplier MSC Supplier No. 48089205
These G-Tek CR seamless knit gloves are designed for cut, puncture, and abrasion resistance, making them suitable for demanding industrial applications. They feature a seamless knit construction for comfort and breathability, and a double-dipped nitrile micro-surface coating on the palm and fingers for enhanced grip in dry and oily conditions.
  • Seamless knit construction offers increased comfort and breathability
  • HPPE blended shell is lightweight, providing excellent dexterity, tactile sensitivity, and cut resistance (ANSI A2)
  • Double-dipped nitrile micro-surface coating provides an elevated grip in dry and oily conditions
  • Coating infused with tiny suction cup pockets that create a vacuum effect, dispensing fluids away on contact
  • Ideal for electronics, glass cutting, sheet metal handling, general duty, parts assembly, and sharp small parts handling