Learn More
MSC PIP Sleeve, Nylon, Universal, Black, Hook and Loop, 1 ea
Catalog No. 5000674904
Encompass
This nylon sleeve provides personal protective equipment, featuring a Hook & Loop closure for a secure fit. It is designed to be cut-resistant for various applications.
  • Made of nylon material
  • Universal sleeve size
  • Black color
  • Hook & loop closure type
  • Overall length 7.0000 in.
  • No thumb hole
  • Cut-resistant
  • ANSI/ISEA cut resistance level A4
Catalog No. 50-006-74904 Supplier MSC Supplier No. 50658236
This nylon sleeve provides personal protective equipment, featuring a Hook & Loop closure for a secure fit. It is designed to be cut-resistant for various applications.
  • Made of nylon material
  • Universal sleeve size
  • Black color
  • Hook & loop closure type
  • Overall length 7.0000 in.
  • No thumb hole
  • Cut-resistant
  • ANSI/ISEA cut resistance level A4