Learn More
Echelon Biosciences Research Labs PIP STRIPS - 10 pack
SDP
Catalog No. 50143989
Encompass
PIP Strips are 2 x 6 cm hydrophobic membranes that have been spotted with 100 pmol of all eight phosphoinositides and seven other biological important lipids. They are used to determine how your protein of interest interacts with one or more of these lipids in a simple protein-lipid overlay assay. Bound protein is then detected by an antibody against the protein or affinity tag. This item is non returnable
This item is not returnable. View return policy
Catalog No. 50-143-989 Supplier Echelon Biosciences Research Labs Supplier No. P600110PK
PIP Strips are 2 x 6 cm hydrophobic membranes that have been spotted with 100 pmol of all eight phosphoinositides and seven other biological important lipids. They are used to determine how your protein of interest interacts with one or more of these lipids in a simple protein-lipid overlay assay. Bound protein is then detected by an antibody against the protein or affinity tag. This item is non returnable