Learn More
MSC PIP Surveyors Tech Vest, ANSI Class 2, Large, 1 ea
Catalog No. 5000673926
Encompass
This tech vest combines a breathable mesh top with a durable solid polyester bottom, offering comfort and resilience. Designed with multiple pockets for organized storage, it features 2-inch retroreflective two-tone tape, meeting ANSI/ISEA 107-2020 Type R Class 2 standards for high visibility in hazardous environments.
  • Breathable mesh top
  • Durable solid polyester bottom
  • Two external flap pockets (sized to fit tablets)
  • Two internal pockets
  • Two pen pockets (one internal & one external)
  • One radio/cell pocket
  • One ID pocket
  • One large back pocket with zipper closure
  • Retroreflective two-tone tape, 2 in
  • Meets ANSI/ISEA 107-2020 Type R Class 2 standards
Catalog No. 50-006-73926 Supplier MSC Supplier No. 68527993
This tech vest combines a breathable mesh top with a durable solid polyester bottom, offering comfort and resilience. Designed with multiple pockets for organized storage, it features 2-inch retroreflective two-tone tape, meeting ANSI/ISEA 107-2020 Type R Class 2 standards for high visibility in hazardous environments.
  • Breathable mesh top
  • Durable solid polyester bottom
  • Two external flap pockets (sized to fit tablets)
  • Two internal pockets
  • Two pen pockets (one internal & one external)
  • One radio/cell pocket
  • One ID pocket
  • One large back pocket with zipper closure
  • Retroreflective two-tone tape, 2 in
  • Meets ANSI/ISEA 107-2020 Type R Class 2 standards