Learn More
Integra Biosciences Corp Pip-tubing connector pack of 5
SDP
Catalog No. 5000881912
Encompass
Pipette-tubing connector pack of 5 for 2 - 6 mm ID silicone tubing
Catalog No. 50-008-81912 Supplier Integra Biosciences Corp Supplier No. 171077
Pipette-tubing connector pack of 5 for 2 - 6 mm ID silicone tubing