Learn More
MSC PIP Welding Helmet, Auto-Darkening, Shade 10, Black, Nylon
Catalog No. 5000507980
Encompass
This auto-darkening welding helmet provides reliable protection and clear visibility for welding applications. It features a durable nylon construction and a solar-powered auto-darkening lens.
  • Auto-darkening lens for enhanced safety and convenience
  • Durable nylon helmet material is breathable, lightweight, and heat resistant
  • Fixed front window style
  • Lens shade of 10
  • Solar power source
  • Green and clear lens colors
  • Black helmet color
Catalog No. 50-005-07980 Supplier MSC Supplier No. 61503926
This auto-darkening welding helmet provides reliable protection and clear visibility for welding applications. It features a durable nylon construction and a solar-powered auto-darkening lens.
  • Auto-darkening lens for enhanced safety and convenience
  • Durable nylon helmet material is breathable, lightweight, and heat resistant
  • Fixed front window style
  • Lens shade of 10
  • Solar power source
  • Green and clear lens colors
  • Black helmet color