Learn More
MSC PIP Welding Helmet, Auto-Darkening, Shade 9 to 13, Nylon, Black
Catalog No. 5000509869
Encompass
This welding helmet features an auto-darkening lens with a shade range of 9 to 13, designed for various welding applications. Constructed from black nylon, it offers breathability, light weight, and heat resistance for user comfort and protection.
  • Auto-darkening lens type
  • Adjustable lens shade from 9 to 13
  • Durable nylon helmet material
  • Black helmet color
  • Solar power source
  • Green and clear lens options
  • Made from a matrix of synthetic polymers
  • Breathable, lightweight, and heat resistant
Catalog No. 50-005-09869 Supplier MSC Supplier No. 61503918
This welding helmet features an auto-darkening lens with a shade range of 9 to 13, designed for various welding applications. Constructed from black nylon, it offers breathability, light weight, and heat resistance for user comfort and protection.
  • Auto-darkening lens type
  • Adjustable lens shade from 9 to 13
  • Durable nylon helmet material
  • Black helmet color
  • Solar power source
  • Green and clear lens options
  • Made from a matrix of synthetic polymers
  • Breathable, lightweight, and heat resistant