Learn More
MSC PIP Welding Jacket, 9 oz Cotton, FR, Black, 1 ea
Catalog No. 5000781320
Encompass
The Ironcat 7050 FR Sateen Cotton Jacket is designed for welding and other industrial applications. Made from flame-resistant sateen cotton, this jacket provides protection and comfort with practical features.
  • Nine oz sateen fabric
  • Anodized snaps and rivets with leather reinforcement
  • Pop-up welders collar with snap closure
  • Inside pocket
  • Scribe/soapstone pocket on left sleeve
  • Made from 100% sateen cotton FR treated material
Catalog No. 50-007-81320 Supplier MSC Supplier No. 21005384
The Ironcat 7050 FR Sateen Cotton Jacket is designed for welding and other industrial applications. Made from flame-resistant sateen cotton, this jacket provides protection and comfort with practical features.
  • Nine oz sateen fabric
  • Anodized snaps and rivets with leather reinforcement
  • Pop-up welders collar with snap closure
  • Inside pocket
  • Scribe/soapstone pocket on left sleeve
  • Made from 100% sateen cotton FR treated material