Learn More
MSC PIP Work Glove, Nylon Blend, Polyurethane Palm, XL, 1 ea
Catalog No. 5000688003
Encompass
This seamless knit glove features a nylon/polyester blend liner and a polyurethane coated flat grip on the palm and fingers. It is designed for various applications, offering comfort and dexterity while being touchscreen compatible.
  • Nylon/polyester blend liner
  • Polyurethane coated flat grip on palm and fingers
  • Touchscreen compatible
  • Knit wrist cuff
  • Coated seamless knit construction
  • Puncture protection level 2 (ASTM F1342)
  • Abrasion protection level 3 (ASTM D3389-10)
  • EN 388 2016 rated: abrasion 4, cut 1, tear 3, puncture 1
Catalog No. 50-006-88003 Supplier MSC Supplier No. 87309571
This seamless knit glove features a nylon/polyester blend liner and a polyurethane coated flat grip on the palm and fingers. It is designed for various applications, offering comfort and dexterity while being touchscreen compatible.
  • Nylon/polyester blend liner
  • Polyurethane coated flat grip on palm and fingers
  • Touchscreen compatible
  • Knit wrist cuff
  • Coated seamless knit construction
  • Puncture protection level 2 (ASTM F1342)
  • Abrasion protection level 3 (ASTM D3389-10)
  • EN 388 2016 rated: abrasion 4, cut 1, tear 3, puncture 1