Learn More
MSC PIP Work Gloves, Nitrile-Coated, Nylon, Seamless Knit, X-Small, 1 Pair
Catalog No. 5000794373
Encompass
These general purpose work gloves feature a seamless knit nylon shell and a nitrile coating on the palm and fingers, making them ideal for manufacturing and assembly applications requiring abrasion, cut, and puncture resistance.
  • Seamless knit nylon shell for increased comfort, finger dexterity, and breathability.
  • Solid nitrile coating on palm and fingers provides excellent dry grip and does not absorb liquids.
  • Knit wrist helps prevent dirt and debris from entering.
  • Breathable back for enhanced comfort during use.
  • Nitrile coating offers excellent puncture, tear, and abrasion resistance.
  • Resistant to hydrocarbon-based oils and solvents.
Catalog No. 50-007-94373 Supplier MSC Supplier No. 44715357
These general purpose work gloves feature a seamless knit nylon shell and a nitrile coating on the palm and fingers, making them ideal for manufacturing and assembly applications requiring abrasion, cut, and puncture resistance.
  • Seamless knit nylon shell for increased comfort, finger dexterity, and breathability.
  • Solid nitrile coating on palm and fingers provides excellent dry grip and does not absorb liquids.
  • Knit wrist helps prevent dirt and debris from entering.
  • Breathable back for enhanced comfort during use.
  • Nitrile coating offers excellent puncture, tear, and abrasion resistance.
  • Resistant to hydrocarbon-based oils and solvents.