Learn More
Enzo Life Sciences PIP2 monoclonal antibody (100µg)
SDP
Supplier:  Enzo Life Sciences ADI915062100
Encompass_Preferred
Clone: not available. Host: Mouse. Isotype: IgG2b. Immunogen: Native PIP2 from bovine spinal cord. Source: Purified from mouse ascites. Species reactivity: Species independent. Applications: ELISA, ICC. Purity: Protein G affinity purified. Formulation: Liquid. In PBS containing 0.05% sodium azide.
Catalog No. 50-200-7107