Learn More
Enzo Life Sciences PIP2 monoclonal antibody (20µl)
SDP
Supplier:  Enzo Life Sciences ADI915052020
Encompass_Preferred
Clone: KT10. Host: Mouse. Isotype: IgG2b. Immunogen: Native PIP2 from bovine spinal cord. Source: Purified from mouse ascites. Species reactivity: Species independent. Applications: ELISA, ICC. Formulation: Liquid. Ascites.
Catalog No. 50-200-7105