Learn More
Aviva Systems Biology Corp PIP3-E Antibody - middle region (ARP46228_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP46228_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against PIP3-E. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human PIP3-E. Alternate names: IPCEF1, KIAA0403, RP3-402L9.2, PIP3-EApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-53464