Learn More
Aviva Systems Biology Corp PIP4K2A Antibody - middle region (ARP56621_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP56621_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against PIP4K2A. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human PIP4K2A. Alternate names: FLJ13267, PIP5K2A, PIP5KII-alpha, PIP5KIIA, PIPK, PI5P4KAApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-61927