Learn More
Aviva Systems Biology Corp PIP5KL1 Antibody - middle region (ARP53244_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP53244_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against PIP5KL1. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human PIP5KL1. Alternate names: MGC46424, PIPKH, RP11-203J24.5, bA203J24.5ApplicationWB. Species reactivity: Cow, Dog, Horse, Human, Pig, Rabbit
Catalog No. 50-014-53706