Learn More
Medchemexpress LLC HY-100703 5mg Medchemexpress, Pipamperone CAS:1893-33-0 Purity:>98%
SDP
Catalog No. 501931921
Encompass_Preferred
Medchemexpress, HY-100703 5mg Pipamperone CAS:1893-33-0 Pipamperone (Floropipamide; McN-JR 3345; R 3345) is a high-affinity antagonist of 5-HT 2A receptor ( pK i =8.2) and D 4 receptor ( pK i =8.0) and a low-affinity antagonist of D 2 receptor (pK i =6.7) [1] . Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-1921 Supplier Medchemexpress LLC Supplier No. HY1007035MG
Medchemexpress, HY-100703 5mg Pipamperone CAS:1893-33-0 Pipamperone (Floropipamide; McN-JR 3345; R 3345) is a high-affinity antagonist of 5-HT 2A receptor ( pK i =8.2) and D 4 receptor ( pK i =8.0) and a low-affinity antagonist of D 2 receptor (pK i =6.7) [1] . Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.