Learn More
AVANTOR MASTERFLEX (VWR LLC) PIPE ADAPT M1/2FTFTX3/4FTFT 10
Catalog No. 502742364
Encompass
Masterflex Fitting, Polypropylene, Straight, Hosebarb to Thread, 3/4IN ID x 1/2IN NPT(M)10/PK, Avantor
Catalog No. 50-274-2364 Supplier AVANTOR MASTERFLEX (VWR LLC) Supplier No. MFLX4061047
Masterflex Fitting, Polypropylene, Straight, Hosebarb to Thread, 3/4IN ID x 1/2IN NPT(M)10/PK, Avantor