Learn More
AVANTOR MASTERFLEX (VWR LLC) PIPE ADAPTER 10-32X3/32FTFT 10
Catalog No. 502742175
Encompass
Masterflex Fitting, Black Nylon, Straight, Hosebarb to Thread Adapter, 3/32IN ID x 10-32 UNF(M) Taper Thread; 10/PK, Avantor
Catalog No. 50-274-2175 Supplier AVANTOR MASTERFLEX (VWR LLC) Supplier No. MFLX4151704
Masterflex Fitting, Black Nylon, Straight, Hosebarb to Thread Adapter, 3/32IN ID x 10-32 UNF(M) Taper Thread; 10/PK, Avantor