Learn More
AVANTOR MASTERFLEX (VWR LLC) Masterflex Fitting, Nylon, Straight, Hose Barb to Threaded Adapter, 1/4"" ID x 1/4"" NPT(F); 10/PK
Catalog No. NC9071571
Encompass
Barbed fittings are designed for quick and easy assembly without tools. Combine these simple and economical fittings with our flexible tubing for use in your low-pressure applications (generally less than 30 psi).
Catalog No. NC9071571 Supplier AVANTOR MASTERFLEX (VWR LLC) Supplier No. MFLX3170220
Barbed fittings are designed for quick and easy assembly without tools. Combine these simple and economical fittings with our flexible tubing for use in your low-pressure applications (generally less than 30 psi).