Learn More
AVANTOR MASTERFLEX (VWR LLC) PIPE ADAPTER M 3/4X3/4 50/PK
Catalog No. 502742216
Encompass
Masterflex Fitting, Polypropylene, Straight, Hosebarb to Thread, 3/4IN ID x 3/4IN NPT(M)50/PK, Avantor
Catalog No. 50-274-2216 Supplier AVANTOR MASTERFLEX (VWR LLC) Supplier No. MFLX4064051
Masterflex Fitting, Polypropylene, Straight, Hosebarb to Thread, 3/4IN ID x 3/4IN NPT(M)50/PK, Avantor