Learn More
Cole-Parmer Fisher Research Labs TW-6 Labs , Pipe and Cable Locator
Supplier:  Cole-Parmer 2000171
Encompass_Preferred
The well-known, M-Scope model, TW-5 Pipe and Cable Locator has been upgraded in performance and user friendliness to model, TW-6. The new model performs the traditional finding tasks of locating underground metal objects, such as pipes, cables, manhole covers, vaults and valve boxes. Like earlier models, the TW-6 will be primarily used by water departments, telephone companies, electric power companies, gas companies and petroleum pipeline companies to know where to dig, and sometimes more importantly, where not to dig in making repairs or rearranging buried lines. The substantial increase in sensitivity of the TW-6 required that we remove all unnecessary metal parts. They collectively inhibited the tracing distance of the more efficient locator. That is why the new very durable catches holding transmitter and receiver together, as well as the individual carrying handles are now completely non-metallic.
Catalog No. 50-630-860