Learn More
Sklar Instruments Cannula Pipe Cleaners, Sklar, Polypropylene. This product is 1/8 in. wide and has an overall length of 12 inches. PK/3.
SDP
Catalog No. 501436985 Shop All Sklar Instruments Products
Encompass_Preferred
Cannula Pipe Cleaners, Sklar, Polypropylene. Sklar Cannula Pipe Cleaners are typically used to clean secretions out of the inner cannula of a tracheostomy tube. They may be included as part of a tracheostomy care kit. This product is 1/8 in. wide and has an overall length of 12 inches. It is sold in packs of three.
This item is not returnable. View return policy
Catalog No. 50-143-6985 Supplier Sklar Instruments Supplier No. 101470
Cannula Pipe Cleaners, Sklar, Polypropylene. Sklar Cannula Pipe Cleaners are typically used to clean secretions out of the inner cannula of a tracheostomy tube. They may be included as part of a tracheostomy care kit. This product is 1/8 in. wide and has an overall length of 12 inches. It is sold in packs of three.