Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisher Science Education™ Pipe Cleaners
SCI_ED

Catalog No. S04193 Shop All Fisherbrand Products
Change view
Click to view available options
Quantity:
1000/Pk.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S04193 1000/Pk.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S04193
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Commonly used in the science classroom.

Specifications

Color Assorted
For Use With (Application) For science classroom
Quantity 1000/Pk.
Length (English) 12 in.
Length (Metric) 30.48 cm
Product Type Pipe Cleaner
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.