Learn More
Grainger Pipe Hanger 2 Pipe 316SS
Catalog No. 502752441
Encompass_Preferred
The Blucher Pipe Hanger Swivel Loop is a pipe hanger designed for suspending pipes from ceilings or beams. It is made from malleable iron and has a working load limit of 250 lbs. The loop diameter is 1-1/4" and it can accommodate pipe sizes from 1/2" to 1-1/4". The loop swivels 360 degrees for easy pipe installation. This pipe hanger allows suspended pipes to be aligned and provides noise reduction. It meets MSS SP-58 standards and is rated for temperatures up to 250°F. The swivel loop comes with a nut and washer for securing to threaded rods up to 3/8" diameter. It can be used in commercial and industrial settings for hanging pipes carrying water, gas, steam or other fluids.
Catalog No. 50-275-2441 Supplier Grainger Supplier No. 14P015
The Blucher Pipe Hanger Swivel Loop is a pipe hanger designed for suspending pipes from ceilings or beams. It is made from malleable iron and has a working load limit of 250 lbs. The loop diameter is 1-1/4" and it can accommodate pipe sizes from 1/2" to 1-1/4". The loop swivels 360 degrees for easy pipe installation. This pipe hanger allows suspended pipes to be aligned and provides noise reduction. It meets MSS SP-58 standards and is rated for temperatures up to 250°F. The swivel loop comes with a nut and washer for securing to threaded rods up to 3/8" diameter. It can be used in commercial and industrial settings for hanging pipes carrying water, gas, steam or other fluids.