Learn More
Grainger Pipe Hanger 3 Pipe 316SS
Catalog No. 502750125
Encompass_Preferred
The Blucher Pipe Hanger Swivel Loop is a pipe hanger designed for suspending pipes from ceilings or beams. It is made from malleable iron and has a working load limit of 250 lbs. The loop size is 1-1/4" ID x 2" OD. This pipe hanger allows the suspended pipe to swivel and has a ball and socket design to prevent binding. It meets MSS SP-58 and MSS SP-69 standards. The swivel loop has a black powder coat finish to help resist corrosion. This versatile pipe hanger can be used in commercial HVAC, fire protection, industrial, and mechanical applications.
Catalog No. 50-275-0125 Supplier Grainger Supplier No. 14P016
The Blucher Pipe Hanger Swivel Loop is a pipe hanger designed for suspending pipes from ceilings or beams. It is made from malleable iron and has a working load limit of 250 lbs. The loop size is 1-1/4" ID x 2" OD. This pipe hanger allows the suspended pipe to swivel and has a ball and socket design to prevent binding. It meets MSS SP-58 and MSS SP-69 standards. The swivel loop has a black powder coat finish to help resist corrosion. This versatile pipe hanger can be used in commercial HVAC, fire protection, industrial, and mechanical applications.