Learn More
Grainger Pipe Hanger 4 Pipe 316SS
Catalog No. 502753052
Encompass_Preferred
The Grainger suspended tube and pipe hangers are designed to support pipes and tubes from ceilings or overhead structures. They feature a steel construction with a durable powder coat finish and include a threaded rod for height adjustment. The hangers have a load capacity up to 500 lbs and accommodate pipe sizes from 1/4" to 4". They can be used in commercial and industrial applications for suspending pipes, tubing, conduit, ducts, and cables. The hangers provide secure support while allowing for adjustment and movement. They are available in multiple sizes to fit different pipe diameters.
Catalog No. 50-275-3052 Supplier Grainger Supplier No. 14P017
The Grainger suspended tube and pipe hangers are designed to support pipes and tubes from ceilings or overhead structures. They feature a steel construction with a durable powder coat finish and include a threaded rod for height adjustment. The hangers have a load capacity up to 500 lbs and accommodate pipe sizes from 1/4" to 4". They can be used in commercial and industrial applications for suspending pipes, tubing, conduit, ducts, and cables. The hangers provide secure support while allowing for adjustment and movement. They are available in multiple sizes to fit different pipe diameters.