Learn More
Grainger Pipe Marker 150# Steam 5 in H 8 in W
Catalog No. 502749251
Encompass_Preferred
The Brady 150 Steam Pipe Marker is a self-adhesive vinyl pipe marker that identifies steam pipes. It has a white background with black text that reads "STEAM" in 1.5 inch tall letters. The marker has an operating temperature range of -40°F to 176°F and is made from flexible, conformable vinyl that resists UV light and chemicals. It meets ANSI A13.1 and NFPA 99 standards for pipe marking and has an expected outdoor life of 5 years. The marker comes in a convenient roll format that is 6 inches wide by 150 feet long for efficient application.
Catalog No. 50-274-9251 Supplier Grainger Supplier No. 14H749
The Brady 150 Steam Pipe Marker is a self-adhesive vinyl pipe marker that identifies steam pipes. It has a white background with black text that reads "STEAM" in 1.5 inch tall letters. The marker has an operating temperature range of -40°F to 176°F and is made from flexible, conformable vinyl that resists UV light and chemicals. It meets ANSI A13.1 and NFPA 99 standards for pipe marking and has an expected outdoor life of 5 years. The marker comes in a convenient roll format that is 6 inches wide by 150 feet long for efficient application.