Learn More
Grainger Pipe Marker 4 1/4 in H 32 in W
Catalog No. 502751307
Encompass_Preferred
The Brady 14J030 Pipe Marker is a self-adhesive vinyl pipe marker that can be used to identify pipes, valves, and fittings. It has a flexible vinyl construction and features a glossy finish that resists fading, abrasion, moisture, and chemicals. This pipe marker comes in orange color and measures 1-1/2" height x 48" length. It can wrap around pipes up to 6" OD and conforms to ANSI A13.1 standards for pipe marking. The adhesive backing allows for easy application and the vinyl material is durable for long-lasting pipe identification. This product helps improve safety and compliance by clearly labeling pipes in facilities.
Catalog No. 50-275-1307 Supplier Grainger Supplier No. 14J030
The Brady 14J030 Pipe Marker is a self-adhesive vinyl pipe marker that can be used to identify pipes, valves, and fittings. It has a flexible vinyl construction and features a glossy finish that resists fading, abrasion, moisture, and chemicals. This pipe marker comes in orange color and measures 1-1/2" height x 48" length. It can wrap around pipes up to 6" OD and conforms to ANSI A13.1 standards for pipe marking. The adhesive backing allows for easy application and the vinyl material is durable for long-lasting pipe identification. This product helps improve safety and compliance by clearly labeling pipes in facilities.