Learn More
Grainger Pipe Marker 5 in H 8 in W
Catalog No. 502752992
Encompass_Preferred
The Brady Pipe Marker 14H743 is a yellow vinyl pipe marker that can be used to identify pipes in industrial facilities. This pipe marker has an aggressive adhesive that adheres to PVC, fiberglass, painted metal, and other common pipe materials. It has a useful temperature range of -40°F to 176°F. The marker is 6" tall and has a width of 3". It meets ANSI standards for pipe marking and identification. This versatile marker works for both indoor and outdoor applications.
Catalog No. 50-275-2992 Supplier Grainger Supplier No. 14H743
The Brady Pipe Marker 14H743 is a yellow vinyl pipe marker that can be used to identify pipes in industrial facilities. This pipe marker has an aggressive adhesive that adheres to PVC, fiberglass, painted metal, and other common pipe materials. It has a useful temperature range of -40°F to 176°F. The marker is 6" tall and has a width of 3". It meets ANSI standards for pipe marking and identification. This versatile marker works for both indoor and outdoor applications.