Learn More
Grainger Pipe Marker 60# Steam 26 in H 12 in W
Catalog No. 502751773
Encompass_Preferred
The Brady Pipe Marker 60-Steam is a self-adhesive vinyl pipe marker designed for identifying steam pipes. It features a high-performance acrylic adhesive and flexible vinyl material that resists UV light, chemicals, abrasion, and moisture. This pipe marker has a temperature range of -40°F to 200°F and comes in a high visibility orange color with black text reading "STEAM". It measures 60" long x 1.5" wide and conforms to ANSI/OSHA requirements for pipe labeling. The Brady Pipe Marker 60-Steam provides an easy and durable pipe identification solution for industrial facilities.
Catalog No. 50-275-1773 Supplier Grainger Supplier No. 14H745
The Brady Pipe Marker 60-Steam is a self-adhesive vinyl pipe marker designed for identifying steam pipes. It features a high-performance acrylic adhesive and flexible vinyl material that resists UV light, chemicals, abrasion, and moisture. This pipe marker has a temperature range of -40°F to 200°F and comes in a high visibility orange color with black text reading "STEAM". It measures 60" long x 1.5" wide and conforms to ANSI/OSHA requirements for pipe labeling. The Brady Pipe Marker 60-Steam provides an easy and durable pipe identification solution for industrial facilities.