Learn More
Grainger Pipe Marker Air 5 in H 8 in W
Catalog No. 502749252
Encompass_Preferred
The Brady Pipe Marker Air is a self-adhesive vinyl marker for identifying compressed air lines. It features a white background with bold black "AIR" text and directional arrows. Made of flexible vinyl, it adheres to curved surfaces and measures 1.5" x 9". It meets ANSI/OSHA standards, enhancing safety by preventing misconnections. The vinyl is UV, chemical, oil, and water-resistant, withstanding -40°F to 176°F. Its adhesive backing allows easy application without tools, making it essential for compliant air line identification.
Catalog No. 50-274-9252 Supplier Grainger Supplier No. 14H752
The Brady Pipe Marker Air is a self-adhesive vinyl marker for identifying compressed air lines. It features a white background with bold black "AIR" text and directional arrows. Made of flexible vinyl, it adheres to curved surfaces and measures 1.5" x 9". It meets ANSI/OSHA standards, enhancing safety by preventing misconnections. The vinyl is UV, chemical, oil, and water-resistant, withstanding -40°F to 176°F. Its adhesive backing allows easy application without tools, making it essential for compliant air line identification.