Learn More
Grainger Pipe Marker Anhydrous Ammnia 9in H 8in W
Catalog No. 502751747
Encompass_Preferred
The Brady Pipe Marker for Anhydrous Ammonia is a 7" x 10" vinyl pipe marker with "Anhydrous Ammonia" text printed in black on a yellow background. It has an aggressive adhesive backing for application on pipes and meets ANSI Z535.1 standards for pipe marking. This durable marker is weather and UV resistant and has a temperature range of -40°F to 200°F. It can be used to identify pipes carrying anhydrous ammonia in industrial facilities. The marker comes in packs of 10.
Catalog No. 50-275-1747 Supplier Grainger Supplier No. 14H754
Add to Cart
Add to Cart
The Brady Pipe Marker for Anhydrous Ammonia is a 7" x 10" vinyl pipe marker with "Anhydrous Ammonia" text printed in black on a yellow background. It has an aggressive adhesive backing for application on pipes and meets ANSI Z535.1 standards for pipe marking. This durable marker is weather and UV resistant and has a temperature range of -40°F to 200°F. It can be used to identify pipes carrying anhydrous ammonia in industrial facilities. The marker comes in packs of 10.