Learn More
Grainger Pipe Marker Boiler Blowdown 5in H 8in W
Catalog No. 502753648
Encompass_Preferred
The Brady Pipe Marker Boiler Blowdown is a self-adhesive vinyl pipe marker that identifies boiler blowdown pipes. It has a white background with black text reading "BOILER BLOWDOWN" in 1-1/2 inch tall letters. This pipe marker meets ANSI standards for pipe marking and has a temperature range of -40°F to 176°F. It has an outdoor durability of 5 years. The marker comes in rolls of 50 markers that are 1-1/2 inches wide x 9 inches long. This product helps identify boiler blowdown pipes for safety and compliance.
Catalog No. 50-275-3648 Supplier Grainger Supplier No. 14H764
The Brady Pipe Marker Boiler Blowdown is a self-adhesive vinyl pipe marker that identifies boiler blowdown pipes. It has a white background with black text reading "BOILER BLOWDOWN" in 1-1/2 inch tall letters. This pipe marker meets ANSI standards for pipe marking and has a temperature range of -40°F to 176°F. It has an outdoor durability of 5 years. The marker comes in rolls of 50 markers that are 1-1/2 inches wide x 9 inches long. This product helps identify boiler blowdown pipes for safety and compliance.