Learn More
Grainger Pipe Marker Boiler Blowdown 9in H 8in W
Catalog No. 502753470
Encompass_Preferred
The Brady Pipe Marker for Boiler Blowdown is a self-adhesive vinyl pipe marker that identifies boiler blowdown pipes. It has a white background with black text that reads "BOILER BLOWDOWN" in 1-1/2 inch tall letters. This pipe marker meets ANSI standards for pipe marking and has a temperature range of -40°F to 176°F. It has an outdoor durability of 5 years. The pipe marker comes in rolls that are 1-1/2 inches wide x 50 feet long. It can be applied by hand or with a wrap-around pipe marker applicator. This pipe marker helps identify boiler blowdown pipes for safety and compliance.
Catalog No. 50-275-3470 Supplier Grainger Supplier No. 14H762
The Brady Pipe Marker for Boiler Blowdown is a self-adhesive vinyl pipe marker that identifies boiler blowdown pipes. It has a white background with black text that reads "BOILER BLOWDOWN" in 1-1/2 inch tall letters. This pipe marker meets ANSI standards for pipe marking and has a temperature range of -40°F to 176°F. It has an outdoor durability of 5 years. The pipe marker comes in rolls that are 1-1/2 inches wide x 50 feet long. It can be applied by hand or with a wrap-around pipe marker applicator. This pipe marker helps identify boiler blowdown pipes for safety and compliance.