Learn More
Grainger Pipe Marker Caustic 26 in H 12 in W
Catalog No. 502749358
Encompass_Preferred
The Brady Pipe Marker Caustic is a self-adhesive vinyl pipe marker that identifies caustic piping systems. It has a white background with bold black text reading "CAUSTIC" and meets ANSI/OSHA requirements for pipe labeling. The marker has an operating temperature range of -40°F to 176°F and is made from flexible, conformable vinyl with a permanent acrylic adhesive that adheres to metal, plastic, and other surfaces. It measures 1.5" H x 6" W. This pipe marker from Brady provides clear identification and warning for caustic piping contents.
Catalog No. 50-274-9358 Supplier Grainger Supplier No. 14H771
The Brady Pipe Marker Caustic is a self-adhesive vinyl pipe marker that identifies caustic piping systems. It has a white background with bold black text reading "CAUSTIC" and meets ANSI/OSHA requirements for pipe labeling. The marker has an operating temperature range of -40°F to 176°F and is made from flexible, conformable vinyl with a permanent acrylic adhesive that adheres to metal, plastic, and other surfaces. It measures 1.5" H x 6" W. This pipe marker from Brady provides clear identification and warning for caustic piping contents.