Learn More
Grainger Pipe Marker Chemical Feed 26in H 12in W
Catalog No. 502752305
Encompass_Preferred
The Brady Pipe Marker Chemical Feed is a self-adhesive vinyl pipe marker that identifies chemical feed lines. It has a legend that reads "Chemical Feed" in black text on a yellow background. The marker has an aggressive adhesive that adheres to PVC, ABS, fiberglass, and epoxy piping. It measures 1" high x 9" long and comes in a pack of 10 markers. The markers are weather and UV resistant for indoor and outdoor use. They can withstand temperatures from -40°F to 176°F. The markers comply with ANSI/ASME and OSHA standards for pipe marking and hazard communication.
Catalog No. 50-275-2305 Supplier Grainger Supplier No. 14H775
The Brady Pipe Marker Chemical Feed is a self-adhesive vinyl pipe marker that identifies chemical feed lines. It has a legend that reads "Chemical Feed" in black text on a yellow background. The marker has an aggressive adhesive that adheres to PVC, ABS, fiberglass, and epoxy piping. It measures 1" high x 9" long and comes in a pack of 10 markers. The markers are weather and UV resistant for indoor and outdoor use. They can withstand temperatures from -40°F to 176°F. The markers comply with ANSI/ASME and OSHA standards for pipe marking and hazard communication.