Learn More
Grainger Pipe Marker Chemical Feed 5 in H 8 in W
Catalog No. 502752187
Encompass_Preferred
The Brady Pipe Marker Chemical Feed is a self-adhesive vinyl pipe marker that identifies chemical feed lines. It has a legend that reads "Chemical Feed" in black text on a yellow background. The marker has an aggressive adhesive that adheres to PVC, ABS, fiberglass reinforced plastic, and other low surface energy materials. It measures 1" high x 8" long and comes in a pack of 10 markers. The markers are weather and UV resistant for long outdoor durability. They provide quick, easy identification of chemical feed piping systems.
Catalog No. 50-275-2187 Supplier Grainger Supplier No. 14H776
The Brady Pipe Marker Chemical Feed is a self-adhesive vinyl pipe marker that identifies chemical feed lines. It has a legend that reads "Chemical Feed" in black text on a yellow background. The marker has an aggressive adhesive that adheres to PVC, ABS, fiberglass reinforced plastic, and other low surface energy materials. It measures 1" high x 8" long and comes in a pack of 10 markers. The markers are weather and UV resistant for long outdoor durability. They provide quick, easy identification of chemical feed piping systems.