Learn More
Grainger Pipe Marker Chilled Water 9 in H 8 in W
Catalog No. 502752073
Encompass_Preferred
The Brady Pipe Marker for Chilled Water is a self-adhesive vinyl pipe marker that identifies pipes carrying chilled water. It has a blue background with white text reading "CHILLED WATER" and has an operating temperature range of -40°F to 200°F. The marker meets ANSI A13.1 and NFPA 99 standards for pipe marking and has a 5-year outdoor durability. It comes in various sizes to fit pipe diameters from 0.5" to 36". This product provides clear identification and meets safety standards for pipe marking in facilities with chilled water systems.
Catalog No. 50-275-2073 Supplier Grainger Supplier No. 14H777
The Brady Pipe Marker for Chilled Water is a self-adhesive vinyl pipe marker that identifies pipes carrying chilled water. It has a blue background with white text reading "CHILLED WATER" and has an operating temperature range of -40°F to 200°F. The marker meets ANSI A13.1 and NFPA 99 standards for pipe marking and has a 5-year outdoor durability. It comes in various sizes to fit pipe diameters from 0.5" to 36". This product provides clear identification and meets safety standards for pipe marking in facilities with chilled water systems.