Learn More
Grainger Pipe Marker Chilled Water Supply 26in H
Catalog No. 502752067
Encompass_Preferred
The Brady Pipe Marker for Chilled Water is a self-adhesive vinyl pipe marker that identifies chilled water pipes. It has a blue background with white text that reads "CHILLED WATER" and has an operating temperature range of -40°F to 200°F. The marker measures 1.5" H x 9" W and is made from flexible, conformable vinyl with a permanent acrylic adhesive that adheres to pipes, valves, ductwork and other surfaces. It helps identify pipes and their contents to improve safety and avoid accidental misconnections. This durable marker resists UV light, abrasion, moisture and chemicals. It meets ANSI/OSHA requirements for pipe marking and labeling.
Catalog No. 50-275-2067 Supplier Grainger Supplier No. 14H786
The Brady Pipe Marker for Chilled Water is a self-adhesive vinyl pipe marker that identifies chilled water pipes. It has a blue background with white text that reads "CHILLED WATER" and has an operating temperature range of -40°F to 200°F. The marker measures 1.5" H x 9" W and is made from flexible, conformable vinyl with a permanent acrylic adhesive that adheres to pipes, valves, ductwork and other surfaces. It helps identify pipes and their contents to improve safety and avoid accidental misconnections. This durable marker resists UV light, abrasion, moisture and chemicals. It meets ANSI/OSHA requirements for pipe marking and labeling.