Learn More
Grainger Pipe Marker City Water 4 in H 24 in W
Catalog No. 502752162
Encompass_Preferred
The Brady Pipe Marker City Water is a self-adhesive vinyl pipe marker that identifies city water pipes. It has a blue background with white text that reads "CITY WATER" and has an operating temperature range of -40°F to 176°F. This pipe marker meets ANSI/OSHA requirements for pipe labeling and has a 5 year outdoor durability. It comes in various sizes to fit pipe diameters from 0.5" to 36".
Catalog No. 50-275-2162 Supplier Grainger Supplier No. 14H788
The Brady Pipe Marker City Water is a self-adhesive vinyl pipe marker that identifies city water pipes. It has a blue background with white text that reads "CITY WATER" and has an operating temperature range of -40°F to 176°F. This pipe marker meets ANSI/OSHA requirements for pipe labeling and has a 5 year outdoor durability. It comes in various sizes to fit pipe diameters from 0.5" to 36".