Learn More
Grainger Pipe Marker Compressed Air 26in H 12in W
Catalog No. 502752398
Encompass_Preferred
The Brady Pipe Marker for Compressed Air is a self-adhesive vinyl pipe marker that identifies compressed air lines. It has a blue background with the words "Compressed Air" in white text, and comes in sizes ranging from 1" to 4" width. The marker is made from flexible, conformable vinyl that resists UV light, chemicals, abrasion, and moisture. It can withstand temperatures from -40°F to 176°F. This pipe marker meets ANSI/OSHA requirements for pipe labeling and helps identify compressed air lines for safety and maintenance. It comes in packs of 25 to 100 markers.
Catalog No. 50-275-2398 Supplier Grainger Supplier No. 14H794
The Brady Pipe Marker for Compressed Air is a self-adhesive vinyl pipe marker that identifies compressed air lines. It has a blue background with the words "Compressed Air" in white text, and comes in sizes ranging from 1" to 4" width. The marker is made from flexible, conformable vinyl that resists UV light, chemicals, abrasion, and moisture. It can withstand temperatures from -40°F to 176°F. This pipe marker meets ANSI/OSHA requirements for pipe labeling and helps identify compressed air lines for safety and maintenance. It comes in packs of 25 to 100 markers.