Learn More
Grainger Pipe Marker Compressed Air 5 in H 8 in W
Catalog No. 502751897
Encompass_Preferred
The Brady Pipe Marker for Compressed Air is a self-adhesive vinyl pipe marker that identifies compressed air lines. It has a legend that reads "Compressed Air" in black text on a yellow background. The marker measures 1-1/2" W x 9" L and is made from 6 mil flexible vinyl. It has an acrylic adhesive that adheres to pipes, hoses, valves and fittings. It is suitable for indoor and outdoor use and resists UV rays, abrasion, moisture and chemicals. The marker meets ANSI/OSHA requirements for pipe marking and labeling. It comes in a pack of 10 markers. The Brady Pipe Markers provide quick, easy identification for compressed air lines.
Catalog No. 50-275-1897 Supplier Grainger Supplier No. 14H795
The Brady Pipe Marker for Compressed Air is a self-adhesive vinyl pipe marker that identifies compressed air lines. It has a legend that reads "Compressed Air" in black text on a yellow background. The marker measures 1-1/2" W x 9" L and is made from 6 mil flexible vinyl. It has an acrylic adhesive that adheres to pipes, hoses, valves and fittings. It is suitable for indoor and outdoor use and resists UV rays, abrasion, moisture and chemicals. The marker meets ANSI/OSHA requirements for pipe marking and labeling. It comes in a pack of 10 markers. The Brady Pipe Markers provide quick, easy identification for compressed air lines.