Learn More
Grainger Pipe Marker Compressed Air 9 in H 8 in W
Catalog No. 502751850
Encompass_Preferred
The Brady Pipe Marker for Compressed Air is a self-adhesive vinyl pipe marker that identifies compressed air lines. It has a white background with black text reading "COMPRESSED AIR" and directional flow arrows. The marker measures 1.5" wide x 9" long and is made from flexible, conformable vinyl that adheres to curved surfaces. It has an operating temperature range of -40°F to 176°F and is suitable for indoor and outdoor use. The adhesive backing allows it to stick to pipes, valves, fittings and other surfaces. This pipe marker improves facility safety by clearly identifying compressed air systems. It meets ANSI/OSHA requirements for pipe marking.
Catalog No. 50-275-1850 Supplier Grainger Supplier No. 14H793
The Brady Pipe Marker for Compressed Air is a self-adhesive vinyl pipe marker that identifies compressed air lines. It has a white background with black text reading "COMPRESSED AIR" and directional flow arrows. The marker measures 1.5" wide x 9" long and is made from flexible, conformable vinyl that adheres to curved surfaces. It has an operating temperature range of -40°F to 176°F and is suitable for indoor and outdoor use. The adhesive backing allows it to stick to pipes, valves, fittings and other surfaces. This pipe marker improves facility safety by clearly identifying compressed air systems. It meets ANSI/OSHA requirements for pipe marking.