Learn More
Grainger Pipe Marker Condensate 4 in H 24 in W
Catalog No. 502752474
Encompass_Preferred
The Brady Pipe Marker Condensate is a self-adhesive vinyl pipe marker that identifies condensate lines. It has a white background with black text that reads "CONDENSATE" in 1-inch tall letters. The marker has an aggressive adhesive that adheres to PVC, ABS, CPVC, steel, aluminum, and copper pipes from -40°F to 200°F. It meets ANSI/ASME and NFPA labeling standards. The marker comes in 8-inch lengths and 4-inch widths on a convenient dispenser box for easy application. It provides clear identification and organization of condensate piping systems.
Catalog No. 50-275-2474 Supplier Grainger Supplier No. 14H799
The Brady Pipe Marker Condensate is a self-adhesive vinyl pipe marker that identifies condensate lines. It has a white background with black text that reads "CONDENSATE" in 1-inch tall letters. The marker has an aggressive adhesive that adheres to PVC, ABS, CPVC, steel, aluminum, and copper pipes from -40°F to 200°F. It meets ANSI/ASME and NFPA labeling standards. The marker comes in 8-inch lengths and 4-inch widths on a convenient dispenser box for easy application. It provides clear identification and organization of condensate piping systems.