Learn More
Grainger Pipe Marker Condensate Drain 5in H 8in W
Catalog No. 502752550
Encompass_Preferred
The Brady Pipe Marker Condensate Drain 14H807 is a self-adhesive vinyl marker for identifying condensate drain pipes. It has a white background with bold black text "CONDENSATE DRAIN" for easy visibility. Measuring 1-1/2" H x 9" L, it is made from flexible vinyl that resists UV light and abrasion, withstanding temperatures from -40°F to 176°F. The adhesive backing allows quick application, and the vinyl is durable yet removable. This marker meets ANSI/OSHA requirements and is suitable for HVAC and refrigeration systems.
Catalog No. 50-275-2550 Supplier Grainger Supplier No. 14H807
The Brady Pipe Marker Condensate Drain 14H807 is a self-adhesive vinyl marker for identifying condensate drain pipes. It has a white background with bold black text "CONDENSATE DRAIN" for easy visibility. Measuring 1-1/2" H x 9" L, it is made from flexible vinyl that resists UV light and abrasion, withstanding temperatures from -40°F to 176°F. The adhesive backing allows quick application, and the vinyl is durable yet removable. This marker meets ANSI/OSHA requirements and is suitable for HVAC and refrigeration systems.