Learn More
Grainger Pipe Marker Condenser Water Return 9in H
Catalog No. 502751895
Encompass_Preferred
The Grainger Approved Pipe Marker for Condenser Water is a self-adhesive vinyl pipe marker that identifies condenser water pipes. It has a blue background with white text that reads "Condenser Water". The marker measures 1-1/2" wide x 9" long and is made of durable vinyl that resists abrasion, moisture, and chemicals. It has an easy peel and stick application and can withstand temperatures from -40°F to 200°F. This pipe marker helps identify condenser water pipes for safety and compliance. It meets ANSI/OSHA requirements for pipe marking in industrial facilities.
Catalog No. 50-275-1895 Supplier Grainger Supplier No. 14H813
The Grainger Approved Pipe Marker for Condenser Water is a self-adhesive vinyl pipe marker that identifies condenser water pipes. It has a blue background with white text that reads "Condenser Water". The marker measures 1-1/2" wide x 9" long and is made of durable vinyl that resists abrasion, moisture, and chemicals. It has an easy peel and stick application and can withstand temperatures from -40°F to 200°F. This pipe marker helps identify condenser water pipes for safety and compliance. It meets ANSI/OSHA requirements for pipe marking in industrial facilities.