Learn More
Grainger Pipe Marker Condenser Water Return 4in H
Catalog No. 502752475
Encompass_Preferred
The Brady Pipe Marker Condenser Water is a self-adhesive vinyl pipe marker that identifies condenser water pipes. It has a legend that reads "Condenser Water" in black text on a light blue background. The marker has an aggressive adhesive that adheres to PVC, iron, stainless steel, aluminum, and other pipe materials. It measures 1" high x 9" long and comes in packs of 25. This pipe marker meets ANSI/OSHA requirements for pipe labeling and identification. It provides clear visual identification of condenser water pipes in industrial and commercial facilities. The vinyl material is UV, chemical, abrasion, and temperature resistant for long-lasting pipe identification.
Catalog No. 50-275-2475 Supplier Grainger Supplier No. 14H812
The Brady Pipe Marker Condenser Water is a self-adhesive vinyl pipe marker that identifies condenser water pipes. It has a legend that reads "Condenser Water" in black text on a light blue background. The marker has an aggressive adhesive that adheres to PVC, iron, stainless steel, aluminum, and other pipe materials. It measures 1" high x 9" long and comes in packs of 25. This pipe marker meets ANSI/OSHA requirements for pipe labeling and identification. It provides clear visual identification of condenser water pipes in industrial and commercial facilities. The vinyl material is UV, chemical, abrasion, and temperature resistant for long-lasting pipe identification.