Learn More
Grainger Pipe Marker Fire Protection 4in H 24in W
Catalog No. 502749073
Encompass_Preferred
The Brady Pipe Marker for Fire Protection is a self-adhesive vinyl pipe marker that identifies fire protection piping systems. It has a white background with red text that reads "FIRE PROTECTION" and has an arrow pointing in the direction of flow. The marker meets ANSI/OSHA requirements for pipe labeling and has a temperature range of -40°F to 176°F. It comes in various sizes ranging from 1.5" width x 6" length up to 4" width x 12" length to fit different pipe diameters. This durable and oil-resistant marker provides quick visual identification of fire protection piping.
Catalog No. 50-274-9073 Supplier Grainger Supplier No. 14H841
The Brady Pipe Marker for Fire Protection is a self-adhesive vinyl pipe marker that identifies fire protection piping systems. It has a white background with red text that reads "FIRE PROTECTION" and has an arrow pointing in the direction of flow. The marker meets ANSI/OSHA requirements for pipe labeling and has a temperature range of -40°F to 176°F. It comes in various sizes ranging from 1.5" width x 6" length up to 4" width x 12" length to fit different pipe diameters. This durable and oil-resistant marker provides quick visual identification of fire protection piping.