Learn More
Grainger Pipe Marker Natural Gas 5 in H 8 in W
Catalog No. 502751313
Encompass_Preferred
The Brady Pipe Marker for Natural Gas is a self-adhesive vinyl label designed for identifying natural gas pipes. It features the words "Natural Gas" printed in black on a yellow background. The label measures 5" W x 2" H and is made from flexible, conformable vinyl that adheres well to curved pipe surfaces. It has an outdoor durability of 5 years when used with a Brady label protector. This pipe marker helps promote safety and meets ANSI Z535.1 standards for pipe labeling. It comes in a pack of 10 markers.
Catalog No. 50-275-1313 Supplier Grainger Supplier No. 14H926
The Brady Pipe Marker for Natural Gas is a self-adhesive vinyl label designed for identifying natural gas pipes. It features the words "Natural Gas" printed in black on a yellow background. The label measures 5" W x 2" H and is made from flexible, conformable vinyl that adheres well to curved pipe surfaces. It has an outdoor durability of 5 years when used with a Brady label protector. This pipe marker helps promote safety and meets ANSI Z535.1 standards for pipe labeling. It comes in a pack of 10 markers.