Learn More
Grainger Pipe Marker Nitrogen 26 in H 12 in W
Catalog No. 502750360
Encompass_Preferred
The Brady Pipe Marker Nitrogen is a pre-printed pipe marker that identifies nitrogen gas lines. It is made of durable vinyl with a permanent adhesive backing for easy application. This pipe marker measures 1.5" H x 7" W and features the word "Nitrogen" in black text on a light blue background. It is suitable for indoor and outdoor use and meets ANSI/OSHA requirements for pipe marking. The adhesive backing allows it to adhere to PVC, steel, aluminum, and other pipe materials. This versatile marker provides clear identification and warning for nitrogen gas piping systems.
Catalog No. 50-275-0360 Supplier Grainger Supplier No. 14H933
Add to Cart
Add to Cart
The Brady Pipe Marker Nitrogen is a pre-printed pipe marker that identifies nitrogen gas lines. It is made of durable vinyl with a permanent adhesive backing for easy application. This pipe marker measures 1.5" H x 7" W and features the word "Nitrogen" in black text on a light blue background. It is suitable for indoor and outdoor use and meets ANSI/OSHA requirements for pipe marking. The adhesive backing allows it to adhere to PVC, steel, aluminum, and other pipe materials. This versatile marker provides clear identification and warning for nitrogen gas piping systems.